Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012173.3 | NP_036305.2 | 7, 8 | 1064 | ||
NM_183420.1 | NP_904356.1 | 8, 9 | 1114 | |||
NM_183421.1 | NP_904357.1 | 8, 9 | 1114 | |||
XM_011534748.2 | XP_011533050.1 | 8, 9 | 887 | |||
XM_011534749.1 | XP_011533051.1 | 7, 8 | 824 | |||
XM_017013313.1 | XP_016868802.1 | 8, 9 | 938 | |||
XM_017013314.1 | XP_016868803.1 | 7, 8 | 1016 | |||
GenBank mRNA | AB070270.1 | - | 7, 8 | 848 | ||
AF174605.1 | - | 4, 5 | 590 | |||
AF419858.1 | - | 8, 9 | 1114 | |||
AK302334.1 | - | 9, 10 | 789 | |||
BC020249.2 | - | 6, 7 | 806 | 4652811 | ||
BC050393.1 | - | 8, 9 | 967 | 5769518 |