Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF020833.1 | - | 4 | 202 | ||
AF094850.1 | - | 4 | 220 | |||
AK291320.1 | - | 4 | 221 | |||
AK293374.1 | - | 4 | 305 | |||
AK296377.1 | - | 2 | 851 | |||
AK309944.1 | - | 4 | 219 | |||
AK311624.1 | - | 4 | 354 | |||
AK315896.1 | - | 4 | 226 | |||
AK316040.1 | - | 3 | 350 | |||
AK316335.1 | - | 3 | 356 | |||
BC000733.2 | - | 4 | 221 | 3504640 | ||
BC008469.1 | - | 4 | 222 | 4279770 | ||
BF701069.1 | - | 4 | 239 | |||
BT006889.1 | - | 4 | 179 | |||
CR457134.1 | - | 4 | 179 | |||
DC356593.1 | - | 4 | 244 | |||
U96074.1 | - | 4 | 188 |