Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCAN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251973.1 | NP_001238902.1 | 5 | 1448 | ||
NM_001251974.1 | NP_001238903.1 | 5 | 1401 | |||
NM_005822.3 | NP_005813.2 | 4 | 1523 | |||
XM_011514226.1 | XP_011512528.1 | 5 | 1490 | |||
GenBank mRNA | AK090990.1 | - | 5 | 1372 | ||
AY034085.1 | - | 5 | 1355 | |||
AY034086.1 | - | 5 | 1368 | |||
BC038509.1 | - | 4 | 1306 | 5259147 | ||
D83407.1 | - | 4 | 1299 |