Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005830.3 | NP_005821.2 | 3 | 593 | ||
GenBank mRNA | AK315248.1 | - | 3 | 551 | ||
BC022045.1 | - | 3 | 561 | 4774270 | ||
Z68747.1 | - | 3 | 523 |