Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047160.2 | NP_001040625.1 | 11 | 1455 | ||
NM_005863.4 | NP_005854.2 | 9 | 1366 | |||
NR_073040.1 | 9 | 1399 | ||||
GenBank mRNA | AB209852.1 | - | 11 | 1401 | ||
AJ010046.1 | - | 9 | 1299 | |||
AK024919.1 | - | 9 | 1306 | |||
AK294244.1 | - | 5 | 832 | |||
AK299335.1 | - | 11 | 1411 | |||
AK302687.1 | - | 11 | 1237 | |||
AK304028.1 | - | 9 | 1341 | |||
AK315566.1 | - | 9 | 1308 | |||
BC010285.1 | - | 9 | 1297 | 3048980 | ||
BC053553.1 | - | 11 | 1407 | 6137683 | ||
BX537509.1 | - | 5 | 4004 | |||
U02081.1 | - | 7 | 887 |