Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRVI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098579.2 | NP_001092049.2 | 9 | 1473 | ||
NM_001100163.2 | NP_001093633.1 | 10 | 1751 | |||
NM_001100167.2 | NP_001093637.1 | 10 | 1468 | |||
NM_001206880.1 | NP_001193809.1 | 9 | 1186 | |||
NM_001206881.1 | NP_001193810.1 | 8 | 1593 | |||
NM_130385.3 | NP_569056.4 | 10 | 1804 | |||
GenBank mRNA | AF081249.1 | - | 8 | 1489 | ||
AF081250.1 | - | 8 | 1217 | |||
AK127209.1 | - | 10 | 1446 | |||
AK296336.1 | - | 10 | 1568 | |||
AK300358.1 | - | 9 | 1458 | |||
AK301296.1 | - | 9 | 797 | |||
AK304702.1 | - | 9 | 820 | |||
BC117127.1 | - | 10 | 1341 | 40125678 |