Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPLA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001279356.1 | NP_001266285.1 | 7 | 705 | ||
NM_001279357.1 | NP_001266286.1 | 7 | 759 | |||
NM_001279358.1 | NP_001266287.1 | 8 | 765 | |||
NM_001279359.1 | NP_001266288.1 | 8 | 767 | |||
NM_001279360.1 | NP_001266289.1 | 8 | 749 | |||
NM_006330.3 | NP_006321.1 | 8 | 807 | |||
XM_005251127.4 | XP_005251184.1 | 8 | 607 | |||
XM_017012956.1 | XP_016868445.1 | 7 | 718 | |||
XM_017012957.1 | XP_016868446.1 | 9 | 757 | |||
XM_017012958.1 | XP_016868447.1 | 8 | 642 | |||
XM_017012959.1 | XP_016868448.1 | 9 | 806 | |||
XM_017012960.1 | XP_016868449.1 | 7 | 624 | |||
GenBank mRNA | AF035293.1 | - | 2 | 236 | ||
AF052112.1 | - | 8 | 807 | |||
AF077198.1 | - | 8 | 618 | |||
AF077199.1 | - | 7 | 570 | |||
AF081281.1 | - | 8 | 648 | |||
AF291053.1 | - | 8 | 631 | |||
AK289400.1 | - | 8 | 691 | |||
AK298206.1 | - | 7 | 549 | |||
AK313739.1 | - | 8 | 644 | |||
BC008652.2 | - | 8 | 695 | 3865775 | ||
BC010397.1 | - | 8 | 619 | 4109335 | ||
CR457103.1 | - | 8 | 613 |