Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006332.4 | NP_006323.2 | 2 | 264 | ||
GenBank mRNA | AB049659.1 | - | 2 | 252 | ||
AF097362.1 | - | 2 | 235 | |||
AK123477.1 | - | 1 | 941 | |||
AK291551.1 | - | 2 | 249 | |||
AL558891.3 | - | 2 | 264 | |||
BC021136.1 | - | 2 | 233 | 4863531 | ||
BC031020.1 | - | 2 | 231 | 4690088 | ||
BE515053.1 | - | 2 | 379 | |||
DQ884410.1 | - | 2 | 202 |