Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006332.4 | NP_006323.2 | 7 | 947 | ||
GenBank mRNA | AB049659.1 | - | 7 | 935 | ||
AF401212.1 | - | 1 | 192 | |||
AK123477.1 | - | 5 | 2042 | |||
AK291551.1 | - | 7 | 932 | |||
BC021136.1 | - | 7 | 916 | 4863531 | ||
BC031020.1 | - | 7 | 914 | 4690088 |