Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006351.3 | NP_006342.2 | 4 | 497 | ||
GenBank mRNA | AF026030.1 | - | 4 | 314 | ||
AF041254.1 | - | 4 | 355 | |||
AK223062.1 | - | 4 | 340 | |||
AK289634.1 | - | 4 | 497 | |||
AK309415.1 | - | 5 | 448 | |||
BC033628.1 | - | 4 | 327 | 5248163 | ||
X58234.1 | - | 35 |