Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006423.2 | NP_006414.2 | 5 | 789 | ||
GenBank mRNA | AF112202.1 | - | 5 | 719 | ||
AF112996.1 | - | 5 | 714 | |||
AJ133534.1 | - | 5 | 735 | |||
BC008950.2 | - | 5 | 732 | 2963628 | ||
CF242972.1 | - | 5 | 745 |