Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNLY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302758.1 | NP_001289687.1 | 5 | 619 | ||
NM_006433.4 | NP_006424.2 | 4 | 538 | |||
NM_012483.3 | NP_036615.2 | 5 | 782 | |||
GenBank mRNA | AK296106.1 | - | 2 | 383 | ||
AK304728.1 | - | 5 | 552 | |||
BC023576.2 | - | 4 | 413 | 4766109 | ||
CR541859.1 | - | 4 | 410 | |||
X05044.1 | - | 5 | 642 | |||
X54101.1 | - | 4 | 538 |