Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006442.3 | NP_006433.2 | 2 | 321 | ||
GenBank mRNA | BC010025.2 | - | 2 | 129 | 3029273 | |
BG719065.1 | - | 2 | 324 | |||
X96506.1 | - | 2 | 76 |