Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261464.1 | NP_001248393.1 | 2 | 303 | ||
NM_001261465.1 | NP_001248394.1 | 2 | 273 | |||
NM_001261466.1 | NP_001248395.1 | 3 | 302 | |||
NM_003815.4 | NP_003806.3 | 2 | 273 | |||
NM_207191.2 | NP_997074.1 | 2 | 273 | |||
NM_207194.2 | NP_997077.1 | 2 | 273 | |||
NM_207195.2 | NP_997078.1 | 2 | 273 | |||
NM_207196.2 | NP_997079.1 | 2 | 273 | |||
NM_207197.2 | NP_997080.1 | 2 | 273 | |||
NR_048577.1 | 2 | 273 | ||||
NR_048578.1 | 2 | 273 | ||||
NR_048579.1 | 2 | 273 | ||||
GenBank mRNA | AB209157.1 | - | 1 | 1804 | ||
AK000667.1 | - | 2 | 259 | |||
AK075498.1 | - | 2 | 229 | |||
AK295614.1 | - | 2 | 245 | |||
AK296925.1 | - | 2 | 224 | |||
AK297468.1 | - | 3 | 223 | |||
AK308172.1 | - | 2 | 196 | |||
AY518542.1 | - | 2 | 158 | |||
AY560593.1 | - | 2 | 259 | |||
AY560594.1 | - | 2 | 259 | |||
AY560595.1 | - | 2 | 259 | |||
AY560596.1 | - | 2 | 259 | |||
AY560597.1 | - | 2 | 259 | |||
AY560598.1 | - | 2 | 259 | |||
AY560599.1 | - | 2 | 259 | |||
AY560600.1 | - | 2 | 259 | |||
AY560601.1 | - | 2 | 259 | |||
AY576417.1 | - | 2 | 259 | |||
BC014566.2 | - | 2 | 239 | 2958341 | ||
BT009764.1 | - | 2 | 158 | |||
U41767.1 | - | 2 | 165 | |||
U46005.1 | - | 2 | 158 |