Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CELF2 SIRNA Assays ›
Gene Symbol:
CELF2
Gene Name:
CUGBP, Elav-like family member 2
Gene Aliases:
BRUNOL3, CELF-2, CUG-BP2, CUGBP2, ETR-3, ETR3, NAPOR
Chromosome Location:
Chr.10: 10462515 - 11336675 on Build GRCh38
UniGene ID:
Hs.309288
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
135874
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CELF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025076.2
NP_001020247.1
7
853
NM_001025077.2
NP_001020248.1
7
710
NM_001083591.1
NP_001077060.1
7
853
NM_001326317.1
NP_001313246.1
9
712
NM_001326318.1
NP_001313247.1
9
712
NM_001326319.1
NP_001313248.1
10
750
NM_001326320.1
NP_001313249.1
10
806
NM_001326321.1
NP_001313250.1
9
712
NM_001326323.1
NP_001313252.1
10
806
NM_001326324.1
NP_001313253.1
8
689
NM_001326325.1
NP_001313254.1
9
783
NM_001326326.1
NP_001313255.1
8
708
NM_001326327.1
NP_001313256.1
8
708
NM_001326328.1
NP_001313257.1
9
802
NM_001326329.1
NP_001313258.1
7
742
NM_001326330.1
NP_001313259.1
7
742
NM_001326331.1
NP_001313260.1
7
894
NM_001326332.1
NP_001313261.1
7
894
NM_001326333.1
NP_001313262.1
6
814
NM_001326334.1
NP_001313263.1
8
1079
NM_001326335.1
NP_001313264.1
7
894
NM_001326336.1
NP_001313265.1
7
894
NM_001326337.1
NP_001313266.1
7
894
NM_001326338.1
NP_001313267.1
6
800
NM_001326339.1
NP_001313268.1
6
800
NM_001326340.1
NP_001313269.1
7
806
NM_001326341.1
NP_001313270.1
7
806
NM_001326342.1
NP_001313271.1
7
806
NM_001326343.1
NP_001313272.1
7
806
NM_001326344.1
NP_001313273.1
7
596
NM_001326345.1
NP_001313274.1
7
674
NM_001326346.1
NP_001313275.1
5
839
NM_001326347.1
NP_001313276.1
7
1004
NM_001326348.1
NP_001313277.1
7
1002
NM_001326349.1
NP_001313278.1
7
1002
NM_006561.3
NP_006552.3
7
799
XM_005252354.4
XP_005252411.1
7
889
XM_005252357.4
XP_005252414.1
5
508
XM_005252358.3
XP_005252415.1
5
849
XM_006717373.2
XP_006717436.1
6
929
XM_011519294.2
XP_011517596.1
7
889
XM_011519297.1
XP_011517599.1
7
1012
XM_017015542.1
XP_016871031.1
6
565
XM_017015543.1
XP_016871032.1
7
655
XM_017015544.1
XP_016871033.1
7
655
XM_017015545.1
XP_016871034.1
7
655
XM_017015546.1
XP_016871035.1
7
655
XM_017015547.1
XP_016871036.1
7
889
XM_017015548.1
XP_016871037.1
7
655
XM_017015549.1
XP_016871038.1
7
655
XM_017015550.1
XP_016871039.1
7
1635
XM_017015551.1
XP_016871040.1
7
889
XM_017015552.1
XP_016871041.1
7
1012
XM_017015553.1
XP_016871042.1
7
1012
XM_017015554.1
XP_016871043.1
7
1012
XM_017015555.1
XP_016871044.1
6
565
XM_017015556.1
XP_016871045.1
6
929
XM_017015557.1
XP_016871046.1
8
594
XM_017015558.1
XP_016871047.1
6
581
XM_017015559.1
XP_016871048.1
6
929
XM_017015560.1
XP_016871049.1
6
590
XM_017015561.1
XP_016871050.1
6
917
XM_017015562.1
XP_016871051.1
6
929
XM_017015563.1
XP_016871052.1
6
916
XM_017015564.1
XP_016871053.1
5
505
XM_017015565.1
XP_016871054.1
5
508
XM_017015566.1
XP_016871055.1
5
505
XM_017015567.1
XP_016871056.1
6
645
XM_017015568.1
XP_016871057.1
8
632
XM_017015569.1
XP_016871058.1
6
932
XM_017015570.1
XP_016871059.1
5
508
XM_017015571.1
XP_016871060.1
6
932
XM_017015572.1
XP_016871061.1
6
633
GenBank mRNA
AF036956.1
-
7
769
AF090693.1
-
7
685
AF090694.1
-
7
637
AF432906.1
-
7
705
AK293164.1
-
7
746
AK295511.1
-
5
440
AK297313.1
-
5
630
AK297380.1
-
5
508
AK299548.1
-
7
629
AK299983.1
-
6
838
AK299992.1
-
7
786
AK302681.1
-
4
404
AK304216.1
-
5
515
AK314087.1
-
7
808
AK316354.1
-
7
605
BC036391.1
-
7
771
5260623
BP362205.1
-
5
352
DA819821.1
-
6
523
U69546.1
-
7
586
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
RNA processing
mRNA processing
regulation of heart contraction
nucleotide binding
RNA binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0