Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDCCAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039707.1 | NP_001034796.1 | 5 | 696 | ||
NM_001039708.1 | NP_001034797.1 | 3 | 477 | |||
NM_006643.3 | NP_006634.3 | 4 | 627 | |||
XM_005266050.3 | XP_005266107.1 | 5 | 650 | |||
XM_005266051.3 | XP_005266108.1 | 4 | 541 | |||
XM_011518157.2 | XP_011516459.1 | 4 | 480 | |||
XM_017014218.1 | XP_016869707.1 | 4 | 797 | |||
XM_017014219.1 | XP_016869708.1 | 4 | 797 | |||
GenBank mRNA | AK001296.1 | - | 3 | 256 | ||
AL559847.3 | - | 5 | 630 | |||
AY349357.1 | - | 4 | 317 | |||
BC014515.1 | - | 4 | 494 | 3547130 | ||
BM549484.1 | - | 477 | ||||
BQ723499.1 | - | 484 |