Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCY2F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001522.2 | NP_001513.2 | 4 | 1468 | ||
GenBank mRNA | BC144241.1 | - | 3 | 1263 | ||
L37378.1 | - | 4 | 1491 |