Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006794.3 | NP_006785.1 | 2 | 1169 | ||
GenBank mRNA | AF072693.1 | - | 2 | 1064 | ||
AK314885.1 | - | 2 | 1125 | |||
BC067475.1 | - | 1 | 965 | 6971705 | ||
BC067476.1 | - | 1 | 886 | 6971810 |
Set Membership: |
G-protein coupled receptor |