Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281437.1 | NP_001268366.1 | 2 | 318 | ||
NM_001281438.1 | NP_001268367.1 | 2 | 322 | |||
NM_007002.3 | NP_008933.2 | 2 | 322 | |||
NM_175573.2 | NP_783163.1 | 2 | 318 | |||
XM_005260257.1 | XP_005260314.1 | 3 | 409 | |||
XM_011528503.1 | XP_011526805.1 | 2 | 1157 | |||
XM_017027602.1 | XP_016883091.1 | 2 | 1157 | |||
XM_017027603.1 | XP_016883092.1 | 3 | 409 | |||
XM_017027604.1 | XP_016883093.1 | 2 | 1153 | |||
XM_017027605.1 | XP_016883094.1 | 2 | 1157 | |||
XM_017027606.1 | XP_016883095.1 | 2 | 1157 | |||
XM_017027607.1 | XP_016883096.1 | 2 | 1153 | |||
GenBank mRNA | AK297567.1 | - | 2 | 216 | ||
AK307459.1 | - | 252 | ||||
AL558522.3 | - | 2 | 254 | |||
BC017245.2 | - | 2 | 218 | 4890786 | ||
BG337785.1 | - | 2 | 278 | |||
CR749213.1 | - | 1 | 260 | |||
D64154.1 | - | 2 | 213 | |||
HY029018.1 | - | 3 | 415 | |||
HY178253.1 | - | 2 | 268 |