Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007018.4 | NP_008949.4 | 2 | 274 | ||
XM_005251679.3 | XP_005251736.1 | 4 | 546 | |||
XM_006716940.2 | XP_006717003.1 | 4 | 546 | |||
XM_006716944.3 | XP_006717007.1 | 4 | 546 | |||
XM_011518166.1 | XP_011516468.1 | 4 | 546 | |||
XM_011518167.1 | XP_011516469.1 | 3 | 387 | |||
XM_011518168.1 | XP_011516470.1 | 4 | 546 | |||
XM_011518169.2 | XP_011516471.1 | 4 | 541 | |||
XM_011518170.1 | XP_011516472.1 | 4 | 546 | |||
XM_011518172.2 | XP_011516474.1 | 4 | 546 | |||
XM_017014223.1 | XP_016869712.1 | 4 | 546 | |||
XM_017014224.1 | XP_016869713.1 | 4 | 546 | |||
XM_017014225.1 | XP_016869714.1 | 3 | 363 | |||
XM_017014227.1 | XP_016869716.1 | 3 | 564 | |||
XM_017014228.1 | XP_016869717.1 | 4 | 551 | |||
XM_017014229.1 | XP_016869718.1 | 4 | 546 | |||
XM_017014230.1 | XP_016869719.1 | 4 | 546 | |||
GenBank mRNA | AA287401.1 | - | 3 | 493 | ||
AF513978.1 | - | 2 | 243 | |||
AK307387.1 | - | 4 | 551 | |||
BC089415.1 | - | 4 | 503 | |||
BC137284.1 | - | 2 | 267 | 9021281 | ||
BC137286.1 | - | 2 | 266 | 9021283 | ||
CF142611.1 | - | 4 | 510 |