Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007021.3 | NP_008952.1 | 2 | 1753 | ||
GenBank mRNA | AB022718.1 | - | 2 | 1679 | ||
AK002191.1 | - | 2 | 1680 | |||
AK098476.1 | - | 2 | 1679 | |||
AL136653.1 | - | 2 | 1684 | |||
BC011402.1 | - | 2 | 1634 | 3853931 |