Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KATNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204076.1 | NP_001191005.1 | 4 | 808 | ||
NM_007044.3 | NP_008975.1 | 4 | 808 | |||
XM_005266804.2 | XP_005266861.1 | 4 | 588 | |||
XM_005266805.3 | XP_005266862.1 | 4 | 727 | |||
XM_017010207.1 | XP_016865696.1 | 5 | 898 | |||
XM_017010208.1 | XP_016865697.1 | 4 | 746 | |||
XM_017010209.1 | XP_016865698.1 | 4 | 588 | |||
XM_017010210.1 | XP_016865699.1 | 4 | 593 | |||
GenBank mRNA | AF056022.1 | - | 3 | 504 | ||
AK292090.1 | - | 4 | 718 | |||
BC050428.1 | - | 4 | 645 | 5764501 |