Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMILIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007046.3 | NP_008977.1 | 4 | 2099 | ||
GenBank mRNA | AF088916.1 | - | 4 | 1600 | ||
AL050138.1 | - | 4 | 1530 | |||
BC009947.2 | - | 1 | 294 | |||
BC136279.1 | - | 4 | 1699 | 9020266 | ||
BC142638.1 | - | 4 | 827 | |||
BC142688.1 | - | 4 | 1981 | 40148654 |