Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102426.1 | NP_001095896.1 | 10 | 1779 | ||
XM_005263862.3 | XP_005263919.1 | 10 | 2068 | |||
XM_005263863.4 | XP_005263920.1 | 10 | 1855 | |||
XM_005263864.3 | XP_005263921.1 | 10 | 2020 | |||
XM_011510507.1 | XP_011508809.1 | 12 | 1910 | |||
XM_011510508.2 | XP_011508810.1 | 10 | 1824 | |||
XM_017003198.1 | XP_016858687.1 | 10 | 2093 | |||
XM_017003199.1 | XP_016858688.1 | 10 | 2046 | |||
XM_017003200.1 | XP_016858689.1 | 10 | 2098 | |||
XM_017003201.1 | XP_016858690.1 | 10 | 2089 | |||
XM_017003202.1 | XP_016858691.1 | 10 | 2137 | |||
GenBank mRNA | AB017114.1 | - | 6 | 907 | ||
AB449889.1 | - | 10 | 1648 | |||
AK292826.1 | - | 8 | 1412 | |||
AK300533.1 | - | 10 | 1824 |