Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102426.1 | NP_001095896.1 | 20 | 3547 | ||
XM_005263862.3 | XP_005263919.1 | 20 | 3836 | |||
XM_005263863.4 | XP_005263920.1 | 20 | 3623 | |||
XM_005263864.3 | XP_005263921.1 | 20 | 3788 | |||
XM_011510507.1 | XP_011508809.1 | 22 | 3678 | |||
XM_017003198.1 | XP_016858687.1 | 19 | 3743 | |||
XM_017003199.1 | XP_016858688.1 | 19 | 3696 | |||
GenBank mRNA | AK000748.1 | - | 3 | 438 | ||
AK292826.1 | - | 18 | 3180 |