Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNPO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191028.2 | NP_001177957.2 | 2, 3 | 728 | ||
NM_012470.3 | NP_036602.1 | 2, 3 | 728 | |||
NR_034053.2 | 3, 4 | 889 | ||||
XM_011515989.2 | XP_011514291.1 | 2, 3 | 330 | |||
GenBank mRNA | AF145029.1 | - | 2, 3 | 438 | ||
AJ133769.1 | - | 2, 3 | 325 | |||
AK225999.1 | - | 2, 3 | 699 | |||
AK312580.1 | - | 2, 3 | 455 | |||
BC068530.1 | - | 2, 3 | 456 |