Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317804.1 | NP_001304733.1 | 4 | 381 | ||
NM_001317805.1 | NP_001304734.1 | 3 | 896 | |||
NM_014294.5 | NP_055109.1 | 3 | 464 | |||
GenBank mRNA | AK289397.1 | - | 3 | 324 | ||
AK292777.1 | - | 3 | 248 | |||
AK303411.1 | - | 4 | 309 | |||
BC000687.2 | - | 3 | 286 | 3347823 | ||
BC020283.1 | - | 3 | 279 | |||
BC037738.1 | - | 3 | 376 | 5261490 | ||
BT007359.1 | - | 3 | 204 | |||
CR541654.1 | - | 3 | 204 | |||
CR541739.1 | - | 3 | 204 | |||
X63679.1 | - | 3 | 325 |