Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014909.4 | NP_055724.1 | 7 | 3342 | ||
XM_017021087.1 | XP_016876576.1 | 9 | 2417 | |||
XM_017021088.1 | XP_016876577.1 | 8 | 2374 | |||
XM_017021089.1 | XP_016876578.1 | 8 | 2233 | |||
XM_017021090.1 | XP_016876579.1 | 7 | 2166 | |||
XM_017021091.1 | XP_016876580.1 | 7 | 3842 | |||
GenBank mRNA | AB028959.1 | - | 7 | 2797 | ||
AL832588.2 | - | 7 | 3045 | |||
BF741850.1 | - | 1 | 127 |