Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHSY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014918.4 | NP_055733.2 | 3 | 2476 | ||
XM_006720435.3 | XP_006720498.1 | 3 | 1758 | |||
XM_011521364.2 | XP_011519666.1 | 4 | 2099 | |||
XM_017022011.1 | XP_016877500.1 | 2 | 1532 | |||
GenBank mRNA | AB023207.1 | - | 3 | 2494 | ||
AB071402.1 | - | 3 | 2014 | |||
AK075096.1 | - | 1 | 206 | |||
AK296943.1 | - | 2 | 1253 | |||
AY358529.1 | - | 3 | 2488 | |||
BC046247.1 | - | 3 | 2483 | 5751618 |