Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CARD8 SIRNA Assays ›
Gene Symbol:
CARD8
Gene Name:
caspase recruitment domain family member 8
Gene Aliases:
CARDINAL, DACAR, DAKAR, NDPP, NDPP1, TUCAN
Chromosome Location:
Chr.19: 48203148 - 48256269 on Build GRCh38
UniGene ID:
Hs.446146
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
136680
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CARD8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001184900.1
NP_001171829.1
9
1251
NM_001184901.1
NP_001171830.1
9
1198
NM_014959.3
NP_055774.2
10
1309
NR_033678.1
8
1168
NR_033679.1
8
1210
XM_006723091.3
XP_006723154.1
11
1462
XM_006723092.3
XP_006723155.1
13
2000
XM_006723093.3
XP_006723156.1
11
1783
XM_006723094.3
XP_006723157.1
11
1442
XM_006723095.3
XP_006723158.1
11
1429
XM_006723096.3
XP_006723159.1
12
1851
XM_006723097.3
XP_006723160.1
10
1634
XM_006723098.3
XP_006723161.1
10
1421
XM_006723099.3
XP_006723162.1
9
1307
XM_006723101.2
XP_006723164.1
10
1324
XM_006723102.3
XP_006723165.1
9
1264
XM_006723104.3
XP_006723167.1
10
1321
XM_011526641.2
XP_011524943.1
14
2307
XM_011526642.2
XP_011524944.1
13
2202
XM_011526643.2
XP_011524945.1
12
1889
XM_011526644.2
XP_011524946.1
10
1474
XM_011526646.2
XP_011524948.1
11
1948
XM_011526647.2
XP_011524949.1
10
1837
XM_011526648.2
XP_011524950.1
11
1954
XM_017026482.1
XP_016881971.1
11
1787
XM_017026483.1
XP_016881972.1
11
1740
XM_017026484.1
XP_016881973.1
11
1748
XM_017026485.1
XP_016881974.1
11
1445
XM_017026486.1
XP_016881975.1
9
1304
XM_017026487.1
XP_016881976.1
8
1673
XM_017026488.1
XP_016881977.1
12
1889
XM_017026489.1
XP_016881978.1
11
1474
GenBank mRNA
AB023172.1
-
8
1203
AF322184.1
-
8
1203
AF331519.1
-
6
890
AF405558.1
-
6
890
AK297069.1
-
9
1188
AK302327.1
-
8
1138
AY026322.1
-
6
890
BX648568.1
-
10
3355
EU118120.1
-
9
1251
EU118121.1
-
8
1210
EU118122.1
-
6
1051
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of interleukin-1 beta secretion
protein binding
cysteine-type endopeptidase activator activity involved in apoptotic process
NACHT domain binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0