Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CARD8 SIRNA Assays ›
Gene Symbol:
CARD8
Gene Name:
caspase recruitment domain family member 8
Gene Aliases:
CARDINAL, DACAR, DAKAR, NDPP, NDPP1, TUCAN
Chromosome Location:
Chr.19: 48203148 - 48256269 on Build GRCh38
UniGene ID:
Hs.446146
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
136681
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CARD8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001184900.1
NP_001171829.1
11
2295
NM_001184901.1
NP_001171830.1
11
2242
NM_001184902.1
NP_001171831.1
13
2516
NM_001184903.1
NP_001171832.1
11
2250
NM_014959.3
NP_055774.2
12
2353
NR_033678.1
10
2212
NR_033679.1
10
2254
NR_033680.1
9
2070
XM_006723091.3
XP_006723154.1
13
2506
XM_006723092.3
XP_006723155.1
15
3044
XM_006723093.3
XP_006723156.1
13
2827
XM_006723094.3
XP_006723157.1
13
2486
XM_006723095.3
XP_006723158.1
13
2473
XM_006723096.3
XP_006723159.1
14
2895
XM_006723097.3
XP_006723160.1
12
2678
XM_006723098.3
XP_006723161.1
12
2465
XM_006723099.3
XP_006723162.1
11
2351
XM_006723101.2
XP_006723164.1
12
2368
XM_006723102.3
XP_006723165.1
11
2308
XM_006723104.3
XP_006723167.1
12
2365
XM_006723106.3
XP_006723169.1
12
2696
XM_006723107.3
XP_006723170.1
11
2221
XM_006723109.3
XP_006723172.1
11
2223
XM_011526641.2
XP_011524943.1
16
3351
XM_011526642.2
XP_011524944.1
15
3246
XM_011526643.2
XP_011524945.1
14
2933
XM_011526644.2
XP_011524946.1
12
2518
XM_011526646.2
XP_011524948.1
13
2992
XM_011526647.2
XP_011524949.1
12
2881
XM_011526648.2
XP_011524950.1
13
2998
XM_011526653.2
XP_011524955.1
12
2886
XM_017026482.1
XP_016881971.1
13
2831
XM_017026483.1
XP_016881972.1
13
2784
XM_017026484.1
XP_016881973.1
13
2792
XM_017026486.1
XP_016881975.1
11
2348
XM_017026487.1
XP_016881976.1
10
2717
XM_017026488.1
XP_016881977.1
14
2933
XM_017026489.1
XP_016881978.1
13
2518
XM_017026492.1
XP_016881981.1
12
2658
XM_017026493.1
XP_016881982.1
11
2550
XM_017026494.1
XP_016881983.1
12
3164
XM_017026495.1
XP_016881984.1
11
2306
XM_017026496.1
XP_016881985.1
11
2226
XM_017026497.1
XP_016881986.1
10
2166
XM_017026498.1
XP_016881987.1
10
2163
XM_017026499.1
XP_016881988.1
9
3936
GenBank mRNA
AB023172.1
-
10
2247
AF322184.1
-
10
2247
AF511652.1
-
9
2105
BX641091.1
-
5
1897
BX648568.1
-
12
4400
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of interleukin-1 beta secretion
protein binding
cysteine-type endopeptidase activator activity involved in apoptotic process
NACHT domain binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0