Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015136.2 | NP_055951.2 | 2 | 192 | ||
XM_005264973.3 | XP_005265030.1 | 2 | 194 | |||
XM_005264974.2 | XP_005265031.1 | 2 | 194 | |||
XM_006713065.1 | XP_006713128.1 | 2 | 194 | |||
XM_017005998.1 | XP_016861487.1 | 2 | 194 | |||
XM_017005999.1 | XP_016861488.1 | 2 | 194 | |||
XM_017006000.1 | XP_016861489.1 | 2 | 194 | |||
XM_017006001.1 | XP_016861490.1 | 2 | 194 | |||
XM_017006002.1 | XP_016861491.1 | 2 | 194 | |||
XM_017006003.1 | XP_016861492.1 | 2 | 194 | |||
XM_017006004.1 | XP_016861493.1 | 2 | 194 | |||
GenBank mRNA | AB052956.1 | - | 2 | 179 | ||
AB052957.1 | - | 2 | 173 | |||
AJ275213.1 | - | 2 | 148 | |||
BC150250.1 | - | 2 | 198 | 9007475 | ||
BE242606.1 | - | 2 | 193 | |||
D87433.2 | - | 2 | 177 | |||
DQ786774.1 | - | 2 | 142 |