Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKS1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015245.2 | NP_056060.2 | 24 | 4159 | ||
XM_005248964.3 | XP_005249021.1 | 25 | 4210 | |||
XM_006715036.2 | XP_006715099.1 | 25 | 4213 | |||
XM_017010593.1 | XP_016866082.1 | 15 | 2636 | |||
GenBank mRNA | AK025381.1 | - | 1 | 47 | ||
BC049213.1 | - | 13 | 2002 | |||
BC132832.1 | - | 24 | 4096 | 40146854 | ||
D86982.1 | - | 24 | 4156 |