Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKS1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015245.2 | NP_056060.2 | 4 | 677 | ||
XM_005248964.3 | XP_005249021.1 | 5 | 728 | |||
XM_006715036.2 | XP_006715099.1 | 5 | 728 | |||
XM_011514431.2 | XP_011512733.1 | 5 | 728 | |||
XM_011514432.2 | XP_011512734.1 | 5 | 728 | |||
XM_011514433.1 | XP_011512735.1 | 5 | 728 | |||
XM_011514434.2 | XP_011512736.1 | 4 | 665 | |||
XM_011514435.2 | XP_011512737.1 | 4 | 665 | |||
XM_011514436.2 | XP_011512738.1 | 5 | 4297 | |||
GenBank mRNA | AK298987.1 | - | 4 | 635 | ||
BC022396.1 | - | 4 | 677 | |||
BC031934.1 | - | 5 | 749 | 4819948 | ||
BC132832.1 | - | 4 | 614 | 40146854 | ||
D86982.1 | - | 4 | 674 |