Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other R3HDM1 SIRNA Assays ›
Gene Symbol:
R3HDM1
Gene Name:
R3H domain containing 1
Gene Aliases:
R3HDM
Chromosome Location:
Chr.2: 135531429 - 135725270 on Build GRCh38
UniGene ID:
Hs.412462
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
136916
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: R3HDM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282798.1
NP_001269727.1
16
2044
NM_001282799.1
NP_001269728.1
13
1652
NM_001282800.1
NP_001269729.1
13
1701
NM_015361.3
NP_056176.2
16
2039
XM_005263626.3
XP_005263683.1
16
1842
XM_005263627.3
XP_005263684.1
16
1842
XM_005263628.3
XP_005263685.1
15
1773
XM_005263629.3
XP_005263686.1
15
1773
XM_006712389.2
XP_006712452.1
16
2028
XM_011510870.1
XP_011509172.1
16
2028
XM_011510871.1
XP_011509173.1
16
2044
XM_011510872.2
XP_011509174.1
15
1986
XM_011510873.2
XP_011509175.1
16
1980
XM_011510874.2
XP_011509176.1
14
1896
XM_011510875.2
XP_011509177.1
14
1912
XM_011510876.2
XP_011509178.1
16
1842
XM_011510877.2
XP_011509179.1
16
1794
XM_011510878.2
XP_011509180.1
15
1773
XM_011510879.2
XP_011509181.1
15
1725
XM_011510881.1
XP_011509183.1
16
2028
XM_017003718.1
XP_016859207.1
16
2044
XM_017003719.1
XP_016859208.1
15
2023
XM_017003720.1
XP_016859209.1
16
1996
XM_017003721.1
XP_016859210.1
16
1858
XM_017003722.1
XP_016859211.1
14
1896
XM_017003723.1
XP_016859212.1
14
1912
XM_017003724.1
XP_016859213.1
14
1731
XM_017003725.1
XP_016859214.1
17
2120
XM_017003726.1
XP_016859215.1
17
2190
XM_017003727.1
XP_016859216.1
16
2148
XM_017003728.1
XP_016859217.1
15
1725
XM_017003729.1
XP_016859218.1
15
1965
XM_017003730.1
XP_016859219.1
13
1709
XM_017003731.1
XP_016859220.1
17
2141
XM_017003732.1
XP_016859221.1
17
2119
XM_017003733.1
XP_016859222.1
17
2163
XM_017003734.1
XP_016859223.1
18
2323
XM_017003735.1
XP_016859224.1
15
1921
XM_017003736.1
XP_016859225.1
15
1853
GenBank mRNA
AK127793.1
-
13
1672
AK290015.1
-
16
1930
AK297000.1
-
8
822
BC041093.1
-
16
1981
5556247
D21852.1
-
13
1606
Back To Top
More Information
Gene Ontology Categories:
Process(es)
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0