Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015836.3 | NP_056651.1 | 6 | 2703 | ||
NM_201263.2 | NP_957715.1 | 6 | 2732 | |||
XM_005270350.3 | XP_005270407.1 | 6 | 2789 | |||
XM_011540494.2 | XP_011538796.1 | 7 | 2772 | |||
XM_011540495.2 | XP_011538797.1 | 5 | 2474 | |||
XM_017000038.1 | XP_016855527.1 | 5 | 2638 | |||
XM_017000039.1 | XP_016855528.1 | 8 | 2958 | |||
XM_017000040.1 | XP_016855529.1 | 6 | 2525 | |||
XM_017000041.1 | XP_016855530.1 | 5 | 2615 | |||
XM_017000042.1 | XP_016855531.1 | 5 | 2609 | |||
GenBank mRNA | BC039889.1 | - | 6 | 2708 | ||
BC044575.1 | - | 6 | 2686 | 5287131 |