Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM114A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317993.1 | NP_001304922.1 | 7 | 748 | ||
NM_001317994.1 | NP_001304923.1 | 6 | 1265 | |||
NM_001317995.1 | NP_001304924.1 | 5 | 491 | |||
NM_018691.3 | NP_061161.2 | 6 | 701 | |||
XM_005268360.4 | XP_005268417.1 | 7 | 826 | |||
XM_005268361.4 | XP_005268418.1 | 6 | 729 | |||
XM_011537540.2 | XP_011535842.1 | 6 | 1055 | |||
XM_011537541.2 | XP_011535843.1 | 6 | 1637 | |||
XM_011537542.2 | XP_011535844.1 | 6 | 926 | |||
XM_017008967.1 | XP_016864456.1 | 6 | 720 | |||
XM_017008968.1 | XP_016864457.1 | 6 | 742 | |||
GenBank mRNA | AF159700.1 | - | 6 | 1194 | ||
AK024664.1 | - | 6 | 666 | |||
AK095446.1 | - | 6 | 621 | |||
AK225821.1 | - | 6 | 684 | |||
AK298943.1 | - | 5 | 460 | |||
AK303865.1 | - | 5 | 541 | |||
AK313331.1 | - | 6 | 673 | |||
BC011414.1 | - | 7 | 683 | 3855873 | ||
BC050701.1 | - | 6 | 662 |