Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASTN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184734.1 | NP_001171663.1 | 2 | 424 | ||
NM_001184735.1 | NP_001171664.1 | 2 | 424 | |||
NM_014010.4 | NP_054729.3 | 16 | 2980 | |||
NM_198186.3 | NP_937829.3 | 2 | 745 | |||
NM_198187.3 | NP_937830.3 | 2 | 424 | |||
NM_198188.2 | NP_937831.1 | 2 | 424 | |||
GenBank mRNA | AB014534.1 | - | 16 | 2980 | ||
AF116574.1 | - | 14 | 2201 | |||
AK024064.1 | - | 2 | 395 | |||
AK127350.1 | - | 15 | 2201 | |||
AK315025.1 | - | 14 | 2044 | |||
BC018759.2 | - | 10 | 1429 | |||
BC029272.1 | - | 2 | 745 | |||
BC093835.1 | - | 2 | 258 | |||
BC101667.1 | - | 2 | 257 | |||
BC143289.1 | - | 2 | 258 | |||
BC143292.1 | - | 2 | 258 | |||
BC143293.1 | - | 2 | 258 | |||
BC146756.1 | - | 16 | 2980 | 9007192 | ||
BM552790.1 | - | 2 | 407 | |||
BX649176.1 | - | 16 | 2315 | |||
DA015423.1 | - | 2 | 424 |