Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABPC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135653.1 | NP_001129125.1 | 7, 8 | 1878 | ||
NM_001135654.1 | NP_001129126.1 | 7, 8 | 1878 | |||
NM_003819.3 | NP_003810.1 | 7, 8 | 1878 | |||
GenBank mRNA | AB208962.1 | - | 1, 2 | 74 | ||
AK222917.1 | - | 1, 2 | 115 | |||
BC029969.1 | - | 7, 8 | 1430 | |||
BC065540.1 | - | 7, 8 | 1781 | 6061561 | ||
BC071591.1 | - | 7, 8 | 1816 | 5265829 | ||
BC094755.1 | - | 7, 8 | 1819 | 30331896 | ||
BC106908.1 | - | 2, 3 | 258 | |||
BC118568.1 | - | 2, 3 | 258 | |||
D12799.1 | - | 4, 5 | 413 | |||
U33818.1 | - | 7, 8 | 1133 | |||
U75686.1 | - | 7, 8 | 1774 |