Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIOK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003831.3 | NP_003822.2 | 3 | 910 | ||
XM_011526242.1 | XP_011524544.1 | 3 | 910 | |||
XM_011526243.1 | XP_011524545.1 | 3 | 910 | |||
GenBank mRNA | AF013591.1 | - | 3 | 386 | ||
AK057292.1 | - | 3 | 910 | |||
AK303938.1 | - | 3 | 398 | |||
BC039729.1 | - | 3 | 425 | 5745005 |