Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYRK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282285.1 | NP_001269214.1 | 4 | 575 | ||
NM_001282286.1 | NP_001269215.1 | 4 | 575 | |||
NM_003845.2 | NP_003836.1 | 12 | 1471 | |||
GenBank mRNA | AF263541.1 | - | 11 | 1371 | ||
AK292322.1 | - | 12 | 1465 | |||
AK298979.1 | - | 3 | 392 | |||
AK303744.1 | - | 4 | 543 | |||
AL561586.3 | - | 4 | 575 | |||
BC031244.1 | - | 12 | 1465 | 5266287 | ||
GU139739.1 | - | 2 | 282 | |||
GU139740.1 | - | 3 | 446 | |||
GU139741.1 | - | 4 | 507 | |||
GU139742.1 | - | 4 | 488 | |||
GU139743.1 | - | 11 | 1455 | |||
GU139744.1 | - | 3 | 445 | |||
GU139745.1 | - | 1141 | ||||
Y09305.1 | - | 4 | 469 |