Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184795.1 | NP_001171724.1 | 4 | 457 | ||
NM_003846.2 | NP_003837.1 | 4 | 651 | |||
NR_073491.1 | 4 | 647 | ||||
NR_073492.1 | 4 | 641 | ||||
NR_073493.2 | 4 | 863 | ||||
GenBank mRNA | AB018080.1 | - | 4 | 433 | ||
AF093670.1 | - | 4 | 431 | |||
AK023991.1 | - | 4 | 415 | |||
AK094173.1 | - | 1 | 1251 | |||
AK301983.1 | - | 4 | 417 | |||
BC011963.1 | - | 4 | 431 | 3866690 | ||
BG423552.1 | - | 391 | ||||
CB155824.1 | - | 4 | 457 | |||
CR542047.1 | - | 4 | 415 | |||
HY108328.1 | - | 4 | 434 |