Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161504.1 | NP_001154976.1 | 15 | 3107 | ||
NM_001319218.1 | NP_001306147.1 | 14 | 3094 | |||
NM_003748.3 | NP_003739.2 | 15 | 3247 | |||
NM_170726.2 | NP_733844.1 | 16 | 2234 | |||
GenBank mRNA | BC007581.1 | - | 15 | 2977 | 3139944 | |
BC023600.2 | - | 15 | 2988 | 4548787 | ||
BX649081.1 | - | 2 | 2350 | |||
FJ462711.1 | - | 14 | 2832 |