Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004274.4 | NP_004265.3 | 2 | 251 | ||
XM_017021808.1 | XP_016877297.1 | 1 | 115 | |||
GenBank mRNA | AB002309.2 | - | 2 | 191 | ||
BC137232.1 | - | 2 | 134 | 9021229 | ||
BC137233.1 | - | 2 | 134 | 9021230 | ||
BC150185.1 | - | 2 | 180 | |||
BC150288.1 | - | 2 | 191 | 8860126 | ||
BC154413.1 | - | 2 | 180 | |||
BP230135.1 | - | 2 | 251 | |||
U17195.2 | - | 2 | 207 |