Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083585.2 | NP_001077054.1 | 4 | 687 | ||
NM_001291581.1 | NP_001278510.1 | 3 | 558 | |||
NM_001291582.1 | NP_001278511.1 | 4 | 687 | |||
NM_004703.5 | NP_004694.2 | 4 | 687 | |||
XM_011524060.1 | XP_011522362.1 | 4 | 687 | |||
XM_017025294.1 | XP_016880783.1 | 3 | 393 | |||
XM_017025295.1 | XP_016880784.1 | 3 | 393 | |||
GenBank mRNA | AF098638.1 | - | 4 | 463 | ||
AK297535.1 | - | 3 | 547 | |||
AK310932.1 | - | 3 | 499 | |||
AK314183.1 | - | 4 | 678 | |||
BC032435.1 | - | 4 | 662 | |||
BC041700.1 | - | 4 | 653 | 6046320 | ||
BI666786.1 | - | 3 | 542 | |||
X77723.1 | - | 4 | 536 | |||
X91141.1 | - | 4 | 651 |