Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLIT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289135.2 | NP_001276064.1 | 1, 2 | 2089 | ||
NM_001289136.2 | NP_001276065.1 | 1, 2 | 2089 | |||
NM_004787.3 | NP_004778.1 | 1, 2 | 2089 | |||
XM_005248211.3 | XP_005248268.1 | 1, 2 | 2089 | |||
XM_006713986.3 | XP_006714049.1 | 1, 2 | 2089 | |||
GenBank mRNA | AB017168.1 | - | 1, 2 | 382 | ||
AF055585.1 | - | 1, 2 | 734 | |||
AF133270.1 | - | 1, 2 | 178 | |||
BC117190.1 | - | 1, 2 | 257 | 40125741 | ||
BC143978.1 | - | 1, 2 | 257 | 9052496 |