Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004855.4 | NP_004846.4 | 12 | 1914 | ||
XM_005254795.4 | XP_005254852.1 | 10 | 1343 | |||
XM_011522235.2 | XP_011520537.1 | 12 | 1869 | |||
XM_011522236.2 | XP_011520538.1 | 11 | 1809 | |||
XM_011522237.2 | XP_011520539.1 | 11 | 1418 | |||
XM_017022730.1 | XP_016878219.1 | 11 | 1333 | |||
XM_017022731.1 | XP_016878220.1 | 12 | 1462 | |||
XM_017022732.1 | XP_016878221.1 | 10 | 1298 | |||
GenBank mRNA | AK093138.1 | - | 3 | 1655 | ||
AK223330.1 | - | 12 | 1619 | |||
AK293666.1 | - | 9 | 1530 | |||
AK314706.1 | - | 12 | 1889 | |||
BC017711.1 | - | 12 | 1851 | 4430974 | ||
D42138.1 | - | 12 | 1642 |
Gene Symbol: CCPG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA740595.1 | - | 1 | 144 |