Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOBEC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270411.1 | NP_001257340.1 | 2 | 132 | ||
NM_004900.4 | NP_004891.4 | 2 | 132 | |||
GenBank mRNA | AK024854.1 | - | 2 | 131 | ||
AU098564.1 | - | 2 | 132 | |||
AY743217.1 | - | 2 | 77 | |||
BC053859.1 | - | 2 | 158 | 6174285 | ||
CT841510.1 | - | 2 | 102 | |||
KF651167.1 | - | 2 | 77 | |||
U61084.1 | - | 2 | 106 |
Gene Symbol: APOBEC3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152426.3 | NP_689639.2 | 2 | 484 | ||
XM_005261337.3 | XP_005261394.1 | 2 | 484 | |||
XM_017028596.1 | XP_016884085.1 | 2 | 484 | |||
GenBank mRNA | BE888971.1 | - | 2 | 86 | ||
BM917407.1 | - | 270 | ||||
CR456396.1 | - | 2 | 145 | |||
DA287097.1 | - | 2 | 479 | |||
DB121958.1 | - | 2 | 486 | |||
EU527884.1 | - | 2 | 77 |