Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISG15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005101.3 | NP_005092.1 | 2 | 555 | ||
GenBank mRNA | AY168648.1 | - | 1 | 448 | ||
BC009507.2 | - | 2 | 489 | 3545944 | ||
BT007297.1 | - | 1 | 448 | |||
BU161226.1 | - | 2 | 512 | |||
M13755.1 | - | 2 | 523 |