Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184795.1 | NP_001171724.1 | 3, 4 | 424 | ||
NM_003846.2 | NP_003837.1 | 3, 4 | 618 | |||
NR_073491.1 | 3, 4 | 614 | ||||
NR_073492.1 | 3, 4 | 608 | ||||
NR_073493.2 | 3, 4 | 830 | ||||
GenBank mRNA | AB018080.1 | - | 3, 4 | 400 | ||
AF093670.1 | - | 3, 4 | 398 | |||
AK023991.1 | - | 3, 4 | 382 | |||
AK094173.1 | - | 1 | 1218 | |||
AK301983.1 | - | 3, 4 | 384 | |||
BC011963.1 | - | 3, 4 | 398 | 3866690 | ||
BG423552.1 | - | 358 | ||||
CB155824.1 | - | 3, 4 | 424 | |||
CR542047.1 | - | 3, 4 | 382 | |||
HY108328.1 | - | 3, 4 | 401 |